Human Recombinant ACRP30 (from E. coli)
Supplier: ProSci
PRSI40-326EA 185 EUR
PRSI40-326
Human Recombinant ACRP30 (from E. coli)
Proteins and Peptides
The gAcrp30 variant is a naturally occurring globular protein, obtained by proteolytic processing of adiponectin. Adiponectin is produced and secreted exclusively by adipocytes, and is a relatively abundant plasma protein, accounting for up to 0.05% of total serum protein. Like Adiponectin, Acrp30 is capable of decreasing hyperglycemia and reversing insulin resistance. Additionally, gAcrp30 has been shown to be an important factor in promoting fat loss by signaling muscle to absorb and burn Free-Fatty Acids (FFAs). The signaling receptors for adiponectin and gAcrp30 have recently been identified and names AdipoR1 and AdipoR2. AdipoR2 is predominantly expressed in the liver. This naturally occurring variant of human gAcrp30/Adipolean is an 18.1 kDa protein, containing 14 amino acids extra at the N-terminus of human gAcrp30/Adipolean.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.