Human Recombinant IL8 (from E. coli)
Supplier: ProSci
PRSI40-331EA 185 EUR
PRSI40-331
Human Recombinant IL8 (from E. coli)
Proteins and Peptides
IL-8 is a proinflammatory CXC chemokine that can signal through the CXCR1 and CXCR2 receptors. It is secreted by monocytes and endothelial cells. IL-8 chemoattracts and activates neutrophils. Recombinant human IL-8 (monocyte-derived) is an 8.4 kDa protein containing 72 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.