Mouse recombinant MIP1A (from E. coli)
Supplier: ProSci
PRSI40-337EA 193 EUR
PRSI40-337
Mouse recombinant MIP1A (from E. coli)
Both MIP-1 alpha and MIP-1 beta are structurally and functionally related CC chemokines. They participate in the host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells e.g. macrophages, lymphocytes and NK cells. While both MIP-1 alpha and MIP-1 beta exert similar effects on monocytes their effect on lymphocytes differ; with MIP-1 alpha selectively attracting CD8+ lymphocytes and MIP-1 beta selectively attracting CD4+ lymphocytes. Additionally, MIP-1 alpha and MIP-1 beta have also been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. Both human and murine MIP-1 alpha and MIP-1 beta are active on human and murine hematopoietic cells. Recombinant murine MIP-1 alpha is a 7.8 kDa protein containing 69 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.