Human Recombinant PAF-AH (from HEK293 cells)
Supplier: ProSci
PRSI40-128EA 185 EUR
PRSI40-128
Human Recombinant PAF-AH (from HEK293 cells)
Proteins and Peptides
Platelet Activating Factor (PAF) is a biologically active phospholipid, which exerts primarily proinflammatory activities by specifically signaling through G-protein-coupled receptors on platelets, neutrophils, and monocytes. Platelet Activating Factor Acetylhydrolase (PAF-AH) is a secreted protein that mediates PAF activity by specifically catalyzing hydrolysis of the "sn2” ester bond, resulting in the conversion of PAF to the biologically inactive lyso-PAF. PAF-AH can also interact with LDL particles to induce the hydrolysis of LDL associated, oxidized phospholipids, generating lysophosphatidylcholine (lyso-PC) and other lysophospholipids. Recombinant PAF-AH is a 420 amino acid glycoprotein which migrates with an apparent molecular mass of 47-55 kDa by SDS-PAGE analysis.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.