Mouse Recombinant MCP1 (from E. coli)
Supplier: ProSci
PRSI40-338EA 185 EUR
PRSI40-338
Mouse Recombinant MCP1 (from E. coli)
Proteins and Peptides
The MCP proteins belong to the CC chemokine family, and signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross reacts across species. Recombinant murine JE is a 13.8 kDa protein containing 125 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.