Human Recombinant Epiregulin (from E. coli)
Supplier: ProSci
PRSI40-149EA 185 EUR
PRSI40-149
Human Recombinant Epiregulin (from E. coli)
Proteins and Peptides
Epiregulin is an EGF related growth factor that binds specifically to EGFR (ErbB1) and ErbB4, but not ErbB2 or ErbB3. It is expressed mainly in the placenta and peripheral blood leukocytes and in certain carcinomas of the bladder, lung, kidney and colon. Epiregulin stimulates the proliferation of keratinocytes, hepatocytes, fibroblasts and vascular smooth muscle cells. It also inhibits the growth of several tumor-derived epithelial cell lines. Human Epiregulin is initially synthesized as a glycosylated 19.0 kDa transmembrane precursor protein, which is processed by proteolytic cleavage to produce a 6.0 kDa mature secreted sequence. Recombinant human Epiregulin is a 5.6 kDa monomeric protein, containing 50 amino residues, which corresponds to the mature secreted Epiregulin sequence.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.