Mouse recombinant MEC (from E. coli)
Supplier: ProSci
PRSI40-357EA 193 EUR
PRSI40-357
Mouse recombinant MEC (from E. coli)
MEC is a secreted CC chemokine expressed primarily by epithelial cells of the bronchioles, salivary gland, mammary gland and colon. MEC signals through the CCR10 receptor and chemoattracts resting CD4, CD8 T-cells and eosinophils. MEC contains six cysteines including the four highly conserved cysteine residues present in CC chemokines. Recombinant murine MEC is a 12.6 kDa protein containing 111 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.