Human PRRT2 peptide
Supplier: ProSci
PRSI7295PEA 105 EUR
PRSI7295P
Human PRRT2 peptide
Proteins and Peptides
18 amino acids near the center of human PRRT2.
  • Can be used to block antibody activity completely in WB by incubating with equal volume of antibody for 30 min at 37°C.
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide