Human Recombinant MGSA (from E. coli)
Supplier: ProSci
PRSI40-366EA 185 EUR
PRSI40-366
Human Recombinant MGSA (from E. coli)
Proteins and Peptides
All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils. GRO/MGSA also stimulates mitogenesis in certain human melanoma cells. Recombinant human GRO/MGSA is a 7.8 kDa protein consisting of 73 amino acids including the 'ELR' motif common to the CXC chemokine family that bind to CXCR1 or CXCR2.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.