Human Recombinant MIG (from E. coli)
Supplier: ProSci
PRSI40-376EA 185 EUR
PRSI40-376
Human Recombinant MIG (from E. coli)
Proteins and Peptides
MIG, a CXC chemokine, is produced by IFN stimulated monocytes, macrophages and endothelial cells. It signals through the CXCR3 receptor. MIG selectively chemoattracts Th1 lymphocytes, and also exerts other activities including inhibition of tumor growth, angiogenesis, and inhibition of colony formation of hematopoietic progenitors. Human MIG is active on murine cells. Recombinant human MIG is an 11.7 kDa protein containing 103 amino acid residues, including the four highly conserved cysteine residues present in CXC chemokines.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.