Rat Recombinant SDF1 alpha (from E. coli)
Supplier: ProSci
PRSI40-414EA 185 EUR
PRSI40-414
Rat Recombinant SDF1 alpha (from E. coli)
Proteins and Peptides
SDF-1 alpha and beta are stromal derived CXC chemokines, and signal through the CXCR4 receptor. SDF-1alpha and beta chemoattract B and T cells, and have been shown to induce migration of CD34+ stem cells. Additionally, the SDF-1 proteins exert HIV suppressive activity in cells expressing the CXCR4 receptor. Recombinant rat SDF-1α is a 7.9 kDa protein containing 68 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.