Human Recombinant SDF1 beta (from E. coli)
Supplier: ProSci
PRSI40-378EA 185 EUR
PRSI40-378
Human Recombinant SDF1 beta (from E. coli)
Proteins and Peptides
SDF-1 alpha and beta are stromal derived CXC chemokines, and signal through the CXCR4 receptor. SDF-1alpha and beta chemoattract B and T cells, and have been shown to induce migration of CD34+ stem cells. Additionally, the SDF-1 proteins exert HIV suppressive activity in cells expressing the CXCR4 receptor. Recombinant human SDF-1beta is an 8.5 kDa protein containing 72 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.