Human Recombinant GCP-2 (from E. coli)
Supplier: ProSci
PRSI40-394EA 193 EUR
PRSI40-394
Human Recombinant GCP-2 (from E. coli)
Human GCP-2 is a member of the CXC family of chemokines. It also promotes neutrophil chemotaxis and degranulation. Recombinant Human GCP-2 is an 7.9 kDa protein containing 72 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.