Human recombinant Lec (from E. coli)
Supplier: ProSci
PRSI40-396EA 193 EUR
PRSI40-396
Human recombinant Lec (from E. coli)
LEC is a CC chemokine that can signal through the CCR8 and CCR1 receptors. It is expressed in the liver, spleen, and thymus. LEC is chemotactic towards monocytes and lymphocytes but not neutrophils. Recombinant human LEC is an 11.2 kDa protein containing 97 amino acid residues, including the four conserved cysteine residues present in CC chemokines.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.