Rat Recombinant GRO beta (from E. coli)
Supplier: ProSci
PRSI40-409EA 185 EUR
PRSI40-409
Rat Recombinant GRO beta (from E. coli)
Proteins and Peptides
All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils. Recombinant rat GRO-beta/MIP-2 is a 7.9 kDa protein consisting of 73 amino acids including the 'ELR' motif common to the CXC chemokine family that bind to CXCR1 or CXCR2.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.