Human Recombinant WNT-1 (from E. coli)
Supplier: ProSci
PRSI40-216EA 185 EUR
PRSI40-216
Human Recombinant WNT-1 (from E. coli)
Proteins and Peptides
Wnt-1 is a secreted protein that signals through the Frizzled family of cell surface receptors and is required for normal embryonic development. Wnt-1 activation induces a complex signaling cascade that ultimately leads to the increased expression of over fifty genes. An important component of Wnt-1 signaling is the stabilization, and resulting accumulation, of the intracellular signaling protein, beta-catenin. Wnt signaling induces and maintains the transformed phenotype and, in certain embryonic cell lines, supports self renewal in the absence of significant differentiation. Elevated levels of Wnt proteins are associated with tumorigenesis and are present in numerous human breast cancers. Mature human Wnt-1 is a glycosylated protein containing 343 amino acid residues. Recombinant human Wnt-1 is a 38.4 kDa, non-glycosylated protein containing 343 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.