Human Recombinant HCC1 (from E. coli)
Supplier: ProSci
PRSI40-390EA 185 EUR
PRSI40-390
Human Recombinant HCC1 (from E. coli)
Proteins and Peptides
HCC-1 is a CC chemokine that signals through the CCR1 receptor and chemoattracts blood monocytes. It is secreted by various tissues including skeletal muscle, heart, spleen, liver and bone marrow. Recombinant human HCC-1 is an 8.4 kDa protein consisting of 72 amino acids including the four highly conserved residues present in CC chemokines.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.