Mouse Recombinant Egf Prfd (from E. coli)
Supplier: ProSci
PRSI40-450EA 185 EUR
PRSI40-450
Mouse Recombinant Egf Prfd (from E. coli)
Proteins and Peptides
Epidermal Growth Factor (EGF) is a potent growth factor that stimulates the proliferation of various epidermal and epithelial cells. Additionally, EGF has been shown to inhibit gastric secretion, and to be involved in wound healing. EGF signals through a receptor known as c-erbB, which is a class I tyrosine kinase receptor. This receptor also binds with TGF-alpha and VGF (vaccinia virus growth factor).
  • Recombinant rat EGF is a 6.2 kDa globular protein containing 54 amino acid residues including 3 intramolecular disulfide-bonds. Recombinant human EGF is a 6.2 kDa globular protein containing 53 amino acid residues including 3 intramolecular disulfide-bonds. Recombinant murine EGF is a 6.0 kDa globular protein containing 53 amino acid residues, including 3 intramolecular disulfide-bonds.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.