Human recombinant TLR-3 (from HEK293 cells)
Supplier: ProSci
PRSI40-239EA 193 EUR
PRSI40-239
Human recombinant TLR-3 (from HEK293 cells)
TLR-3 is a single-pass type I receptor that binds to and signals the presence of microbial pathogens and double stranded RNA (dsRNA) viruses. Signaling through TLR-3 can promote the NF-κB pathway to initiate innate and adaptive immune responses to bacterial and viral infections, as well as the p53 pathway to trigger apoptosis in cells infected with dsRNA viruses. TLR-3 belongs to a family of structurally-related toll-like receptors (TLRs) containing an N-terminal domain rich in leucine repeats and a C-terminal intracellular Toll/interleukin (IL)-1 (TIL) domain. TLR-3 is expressed primarily in dendritic cells of the placenta and pancreas where it can reside on both sides of the plasma membrane and in the endosomal compartment of the cells. Recombinant human TLR-3 is 77.0 kDa glycoprotein containing 681 residues which comprise the TLR-3 extracellular domain.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.