Human recombinant IL17A (from E. coli)
Supplier: ProSci
PRSI40-263EA 193 EUR
PRSI40-263
Human recombinant IL17A (from E. coli)
Human IL-17A is a homodimer of two 136 amino acid chains, secreted by activated T-cells that act on stromal cells to induce production of proinflammatory and hematopoietic bioactive molecules. Today, IL-17 represents a family of structurally-related cytokines that share a highly conserved C-terminal region but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17A exhibits cross-species bioactivity between human and murine cells. Recombinant human IL-17A is a 31.0 kDa disulfide-linked homodimer of two 136 amino acid polypeptide chains.
  • Murine IL-17A is a disulfide linked homodimer, secreted by activated T-cells that act on stromal cells to induce production of proinflammatory and hematopoietic bioactive molecules. Today, IL-17 represents a family of structurally-related cytokines that share a highly conserved C-terminal region but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17A exhibits cross-species bioactivity between human and murine cells. Recombinant murine IL-17A is a 30.0 kDa disulfide-linked homodimer of two 133 amino acid polypeptide chains.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.