Mouse Recombinant IL17F (from E. coli)
Supplier: ProSci
PRSI40-424EA 185 EUR
PRSI40-424
Mouse Recombinant IL17F (from E. coli)
Proteins and Peptides
IL-17F, a member of the IL-17 family of structurally related cytokines, has been shown to stimulate proliferation and activation of T-cells and PBMCs. IL-17F also regulates cartilage matrix turnover and inhibits angiogenesis. Recombinant murine IL-17F is a disulfide-linked homodimer of 30 kDa, consisting of two 134 amino acid residue chains.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.