Mouse recombinant IL2 (from E. coli)
Supplier: ProSci
PRSI40-431EA 193 EUR
PRSI40-431
Mouse recombinant IL2 (from E. coli)
IL-2 is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions which are essential for the immune response. IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine activated killer cells, monocytes, macrophages and oligodendrocytes. Recombinant murine IL-2 is a 17.2 kDa protein, containing 149 amino acid residues. Recombinant rat IL-2 is a 15.3 kDa protein containing 134 amino acid residues. Recombinant human IL-2 is a 15.5 kDa protein, containing 134 amino acid residues including one intrachain disulfide bond.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.