Mouse recombinant IL3 (from E. coli)
Supplier: ProSci
PRSI40-432EA 193 EUR
PRSI40-432
Mouse recombinant IL3 (from E. coli)
IL-3 is a hematopoietic growth factor that promotes the survival, differentiation and proliferation of committed progenitor cells of the megakaryocyte, granulocyte-macrophage, erythroid, eosinophil, basophil and mast cell lineages. Produced by T cells, mast cells and eosinophils, IL-3 enhances thrombopoieses, phagocytosis, and antibody-mediated cellular cytotoxicity. Its ability to activate monocytes suggests that IL-3 may have additional immunoregulatory roles. Many of the IL-3 activities depend upon co-stimulation with other cytokines. IL-3 is species-specific, variably glycosylated cytokine. Recombinant murine IL-3 is a 15.1 kDa globular protein containing 135 amino acid residues. Recombinant human IL-3 is a 15.0 kDa globular protein containing 133 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.