Mouse Recombinant IL33 (from E. coli)
Supplier: ProSci
PRSI40-429EA 185 EUR
PRSI40-429
Mouse Recombinant IL33 (from E. coli)
Proteins and Peptides
IL-33 is a proinflammatory protein that shares structural and functional characteristics with the IL-1 cytokine family. It binds and signals through the IL-1RL1/ST2 receptor activating NF-kappaB and MAP kinases. IL-33 induces production of TH2 cell related cytokines, including IL-4, IL-5 and IL-13, and exerts multiple inflammation related bioactivities. Recombinant murine IL-33 is a 17.5 kDa protein containing 158 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.