Human Recombinant IL5 (from E. coli)
Supplier: ProSci
PRSI40-248EA 185 EUR
PRSI40-248
Human Recombinant IL5 (from E. coli)
Proteins and Peptides
IL-5 is a hematopoietic growth factor that stimulates the proliferation and activation of eosinophils. Produced by mast cells, T cells, and eosinophils, IL-5 plays an important role in inducing cell-mediated immunity against parasitic infections and certain tumors. Elevated levels of IL-5 lead to Eosinophilia, which may result in the induction of asthma and other allergic diseases. Human and murine IL-5 is cross-species reactive. Recombinant human IL-5 is a 26.0 kDa disulfide-linked homodimeric protein containing two 116 amino acid chains. Recombinant rat IL-5 is a 26.0 kDa disulfide-linked homodimeric protein containing two 113 amino acid chains. Recombinant murine IL-5 is a 26.2 kDa disulfide-linked homodimeric protein containing two 113 amino acid chains.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.