Human Recombinant FGF Acidic (from E. coli)
Supplier: ProSci
PRSI40-159EA 185 EUR
PRSI40-159
Human Recombinant FGF Acidic (from E. coli)
Proteins and Peptides
FGF-acidic is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-acidic is a non-glycosylated heparin binding growth factor that is expressed in the brain, kidney, retina, smooth muscle cells, bone matrix, osteoblasts, astrocytes and endothelial cells. FGF-acidic has the ability to signal through all the FGF receptors. Recombinant murine FGF-acidic is a 15.9 kDa protein consisting of 141 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.