Human Recombinant GMFB (from E. coli)
Supplier: ProSci
PRSI40-508EA 193 EUR
PRSI40-508
Human Recombinant GMFB (from E. coli)
GMF-β a brain-specific protein that belongs to the actin-binding proteins (ADF) structural family. GMF-β appears to play a role in the differentiation, maintenance, and regeneration of the nervous system. It also supports the progression of certain auto-immune diseases, possibly through its ability to induce the production and secretion of various pro-inflammatory cytokines. Recombinant human GMF-β is a 16.5 kDa globular protein containing 141 amino acid residues including one intramolecular disulfide bond.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.