Human Recombinant P16-Ink4A-Tat (from E. coli)
Supplier: ProSci
PRSI40-527EA 185 EUR
PRSI40-527
Human Recombinant P16-Ink4A-Tat (from E. coli)
Proteins and Peptides
p16-INK4a is a nuclear protein that regulates the cell cycle by inhibiting cyclin dependent kinase-4 (CDK4) and CDK6. p16-INK4a inhibits CDK activity by binding to the CDK molecules in a manner that interferes with their ability to interact with cyclin D. This activity has the effect of suppressing tumor formation and growth, and of inducing replicative senescence in various normal cells, including stem cells. The expression of p16-INK4a steadily increases with age and tends to accumulate in stem cell compartments. The deletion, rearrangement, or mutation of the p16-INK4a gene is frequently found in melanomas as well as in certain other types of cancer. P16-INK4a and other transcription factors have been introduced into cells by DNA transfection, viral infection, or microinjection. Protein transduction using TAT fusion proteins represents an alternative methodology for introducing transcription factors and other nuclear proteins into primary as well as transformed cells. Recombinant human p16-INK4a-TAT expressed in E. coli is a 18 kDa protein containing 167 amino-acid residues, including the 156 residues of full-length p16-INK4a and a 13-residue C-terminal TAT peptide (GGYGRKKRRQRRR).
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.