Human Recombinant Gremlin 1 (from CHO cells)
Supplier: ProSci
PRSI40-594EA 185 EUR
PRSI40-594
Human Recombinant Gremlin 1 (from CHO cells)
Proteins and Peptides
Gremlin-1 (isoform-1) belongs to a group of diffusible proteins which bind to ligands of the TGF-β family and regulate their activity by inhibiting their access to signaling receptors. The interplay between TGF-β ligands and their natural antagonists has major biological significance during development processes, in which cellular response can vary considerably depending upon the local concentration of the signaling molecule. Gremlin is highly expressed in the small intestine, fetal brain, and colon and lower expression in brain, prostate, pancreas and skeletal muscle. Gremlin-1 regulates multiple functions in early development by specifically binding to and inhibiting the function of BMP-2, -4, and -7. It also plays a role in carcinogenesis and kidney branching morphogenesis. Recombinant Gremlin-1 is a 13.7 kDa protein containing 160 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.