Human Recombinant FGF10 (from E. coli)
Supplier: ProSci
PRSI40-172EA 185 EUR
PRSI40-172
Human Recombinant FGF10 (from E. coli)
Proteins and Peptides
FGF-10 is a heparin binding growth factor that belongs to the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-10 is most related to KGF/FGF-7 and is expressed during the development and preferentially in adult lungs. It signals through the FGFR 2b. Recombinant human FGF-10 is a 19.3 kDa protein consisting of 170 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.