Human Recombinant GDF3 (from E. coli)
Supplier: ProSci
PRSI40-220EA 185 EUR
PRSI40-220
Human Recombinant GDF3 (from E. coli)
Proteins and Peptides
GDF-3 is a member of the TGF-β superfamily of growth and differentiation factors, and is highly homologous to GDF-9. Unlike most TGF-β family members, GDF-3 and GDF-9 are not disulfide-linked dimers. GDF-3 is expressed in adult bone marrow, spleen, thymus, and adipose tissue. The expression of GDF-3 is upregulated in high-fat-fed wild-type FABP4/aP2 null mice and was associated with obesity, but not with the related hyperglycemia/hyperinsulinemia which characterizes Type 2 diabetes. Recombinant human GDF-3 is a 26.0 kDa non-disulfide-linked homodimer containing two 114 amino acid polypeptide chains.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.