Human Recombinant IL33 (from E. coli)
Supplier: ProSci
PRSI40-273EA 185 EUR
PRSI40-273
Human Recombinant IL33 (from E. coli)
Proteins and Peptides
Human IL-33 is a proinflammatory protein that shares structural and functional characteristics with the IL-1 cytokine family. It binds and signals through the IL-1RL1/ST2 receptor activating NF-kappaB and MAP kinases. IL-33 induces production of TH2 cell related cytokines, including IL-4, IL-5 and IL-13, and exerts multiple inflammation related bioactivities. Recombinant human IL-33 is a 17.9 kDa protein containing 159 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.