Mouse Recombinant IL6 (from E. coli)
Supplier: ProSci
PRSI40-112EA 185 EUR
PRSI40-112
Mouse Recombinant IL6 (from E. coli)
Proteins and Peptides
IL-6 is a pleiotropic cytokine that plays an important role in host defense by regulating immune and inflammatory responses. Produced by T cells, monocytes, fibroblasts, endothelial cells and keratinocytes, IL-6 has diverse biological functions. It stimulates B-cell differentiation and antibody production, synergizes with IL-3 in megakaryocyte development and platelet production, induces expression of hepatic acute-phase proteins, and regulates bone metabolism. IL-6 signals through the IL-6 receptor system that consists of two chains, IL-6R α and gp130. Murine IL-6 is inactive on human cells, while both human and murine are equally active on murine cells. Recombinant human IL-6 is a 20.9 kDa protein containing 184 amino acid residues. Recombinant rat IL-6 is a 21.7 kDa protein containing 187 amino acid residues. Recombinant murine IL-6 is a 21.7 kDa protein containing 187 amino acid residues.
  • Recombinant murine IL-6 is a 21.7 kDa protein containing 187 amino acid residues.