Human Recombinant KGF (from E. coli)
Supplier: ProSci
PRSI40-605EA 320 EUR
PRSI40-605
Human Recombinant KGF (from E. coli)
Proteins and Peptides
Keratinocyte Growth Factor (KGF/FGF-7) is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of variety of tissues, by promoting cellular proliferation and differentiation. KGF/FG-7 is a mitogen factor specific for epithelial cells and keratinocytes and signals through FGFR 2b. KGF/FGF-7 plays a role in kidney and lung development, angiogenesis, and wound healing. Recombinant human KGF/FGF-7 is an 18.9 kDa protein consisting of 163 amino acid residues.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.