Human Recombinant Persephin (from E. coli)
Supplier: ProSci
PRSI40-501EA 185 EUR
PRSI40-501
Human Recombinant Persephin (from E. coli)
Proteins and Peptides
Persephin is a disulfide-linked homodimer neurotrophic factor structurally related to GDNF, Artemin, and Neurturin. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. Persephin signals through a multicomponent receptor system, composed of RET and one of four GFR alpha (alpha1-alpha4) receptors. The GFRalpha4 was first identified in chicken and was later shown to be the preferential binding subunit for Persephin. Persephin promotes the survival of ventral midbrain dompaminergic neurons and motor neurons after sciatic nerve oxotomy, and like GNDF, promotes ureteric bud branching. However, in contrast to GDNF and Neurturin, Persephin does not support survival of peripheral neurons. Recombinant human Persephin is a disulfide-linked homodimer, composed of two 10.3 kDa polypeptide chains (192 total amino acid residues). Each chain contains seven conserved cysteine residues, one of which (Cys 63) is used for inter-chain disulfide bridging and the others are involved in intramolecular ring formation known as the cysteine knot configuration. Human Recombinant protein manufactured under license from the University of Washington patent# US 6,716,600; US 6,692,943; US 6,645,937; US 6,403,335; US 6,232,449; US 6,222,022.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.