Human recombinant MCSF (from E. coli)
Supplier: ProSci
PRSI40-606EA 412 EUR
PRSI40-606
Human recombinant MCSF (from E. coli)
Proteins and Peptides
M-CSF is a potent hematopoietic factor produced by a variety of cells including lymphocytes, monocytes, fibroblasts, endothelial cells, myoblasts and osteoblasts. It is a key regulator of cellular proliferation, differentiation, and survival of blood monocytes, tissue macrophages and their progenitor cells. M-CSF has been shown to play important roles in modulating dermal thickness, and male and female fertility. M-CSF is clinically used in the treatment of infection, malignancies and atherosclerosis. It facilitates hematopoietic recovery after bone marrow transplantation. The human is reactive in murine systems, but the murine molecule exhibits no activity on human cells. Recombinant rat M-CSF is a 18.1 kDa homodimeric protein containing two 155 amino acid polypeptide subunits.
  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.