Human Recombinant Rage1 (from HEK293 cells)
Supplier: ProSci
PRSI96-019EA 395 EUR
PRSI96-019
Human Recombinant Rage1 (from HEK293 cells)
Proteins and Peptides
Advanced glycosylation end product-specific receptor (AGER) is also known as RAGE, and is a single-pass type I membrane protein which contains two Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. AGER is highly expressed in endothelial cells. AGER mediates interactions of advanced glycosylation end products (AGE), and also accumulates in vascular tissue in aging and at an accelerated rate in diabetes, as well as acts as a mediator of both acute and chronic vascular inflammation in conditions such as atherosclerosis and in particular as a complication of diabetes. AGER / RAGE signaling plays an important role in regulating the production/expression of TNF-alpha, oxidative stress, and endothelial dysfunction in type 2 diabetes.