Human Recombinant Cripto (from HEK293 cells)
Supplier: ProSci
PRSI96-202EA 455 EUR
PRSI96-202
Human Recombinant Cripto (from HEK293 cells)
Proteins and Peptides
Teratocarcinoma-derived growth factor 1 (TDGF1) is also known as Cripto-1 growth factor (CRGF), Epidermal growth factor-like cripto protein CR1, CRIPTO, is a cell membrane which contains one EGF-like domain. TDGF1 is preferentially expressed in gastric and colorectal carcinomas than in their normal counterparts. TDGF1 interacts with the activin type-1 receptor ACVR1B. TDGF1 could play a role in the determination of the epiblastic cells that subsequently give rise to the mesoderm.