Human Recombinant Egfr2 (from HEK293 cells)
Supplier: ProSci
PRSI96-259EA 336 EUR
PRSI96-259
Human Recombinant Egfr2 (from HEK293 cells)
Proteins and Peptides
The epidermal growth factor receptor (EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. The epidermal growth factor receptor is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: EGFR (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). Mutations affecting EGFR expression or activity could result in cancer.