Human Recombinant LAIR2 (from HEK293 cells)
Supplier: ProSci
PRSI96-504EA 395 EUR
PRSI96-504
Human Recombinant LAIR2 (from HEK293 cells)
Proteins and Peptides
Leukocyte-associated immunoglobulin-like receptor 2 (LAIR2) is also known as CD antigen CD306. LAIR2 contains one Ig-like C2-type (immunoglobulin-like) domain. LAIR2 is a member of the immunoglobulin superfamily and was identified by its similarity to LAIR1, an inhibitory receptor present on mononuclear leukocytes. LAIR2 / CD306 is thought to be secreted and may help modulate mucosal tolerance.