Human Recombinant PLA2 (from HEK293 cells)
Supplier: ProSci
PRSI96-629EA 480 EUR
PRSI96-629
Human Recombinant PLA2 (from HEK293 cells)
Proteins and Peptides
Phosphatidylcholine 2-acylhydrolase 1B (PLA2G1B) is also known as Phospholipase A2 (PLA2), PLA2A, PPLA2, which belongs to the phospholipase A2 family. PLA2G1B is activated by trypsin cleavage in the duodenum and can also be activated by thrombin or autocatalytically. PLA2 catalyzes the calcium-dependent hydrolysis of the 2-acyl groups in 3-sn-phosphoglycerides, this releases glycerophospholipids and arachidonic acid that serve as the precursors of signal molecules.