Human Recombinant DR6 (from HEK293 cells)
Supplier: ProSci
PRSI96-730EA 455 EUR
PRSI96-730
Human Recombinant DR6 (from HEK293 cells)
Proteins and Peptides
Tumor necrosis factor receptor superfamily member 21 (TNFRSF21) is also known as death receptor 6 (DR6), which is a member of the TNF-receptor superfamily. TNFRSF21 contains one death domain and four TNFR-Cys repeats. TNFRSF21 / DR6 has been shown to activate NF-κB and MAPK8/JNK, and induce cell apoptosis. Through its death domain, this receptor interacts with TRADD protein, which is known to serve as an adaptor that mediates signal transduction of TNF-receptors.