Human Recombinant BMP2 (from E. coli)
Supplier: ProSci
PRSI91-010EA 580 EUR
PRSI91-010
Human Recombinant BMP2 (from E. coli)
Proteins and Peptides
Bone Morphogenetic Protein-2 (BMP-2) is one of the bone-growth regulatory factors that belong to the transforming growth factor-beta (TGF-beta) superfamily of proteins. BMPs are synthesized as large precursor molecules, which are cleaved by proteolytic enzymes. The active form of BMP-2 can consist of a dimer of two identical proteins or a heterodimer of two related bone morphogenetic proteins.
  • This recombinant protein can be used for biological assays. For research use only.