Human Recombinant IFN alpha 2a (from E. coli)
Supplier: ProSci
PRSI91-018EA 295 EUR
PRSI91-018
Human Recombinant IFN alpha 2a (from E. coli)
Proteins and Peptides
At least 23 different variants of IFN- alpha are known. The individual proteins have molecular masses between 19-26 kDa and consist of proteins with lengths of 156-166 and 172 amino acids. All IFN- alpha subtypes possess a common conserved sequence region between amino acid positions 115-151 while the amino-terminal ends are variable. Many IFN- alpha subtypes only differ in their sequences by one or two positions. Naturally occurring variants also include proteins truncated by 10 amino acids at the carboxy-terminal end.
  • This recombinant protein can be used for biological assays. For research use only.