Human recombinant IL36RA (from E. coli)
Supplier: ProSci
PRSI91-057EA 748 EUR
PRSI91-057
Human recombinant IL36RA (from E. coli)
Proteins and Peptides
Human Interleukin-36 Receptor Antagonist (IL-36RN) is a secreted protein which belongs to the Interleukin 1 cytokine family (IL-1 family). IL-36RN is predominantly expressed in keratinocytes but not in fibroblasts, endothelial cells or melanocytes. IL-36RN is also detected in the spleen, brain leukocyte and macrophage cell types. Increased in lesional psoriasis skin. IL-36RN is a highly and a specific antagonist of the IL-1 receptor-related protein 2-mediated response to Interleukin 1 family member 9 (IL1F9). Dysregulated expression of novel agonistic and antagonistic IL-1 family member ligands can promote cutaneous inflammation, revealing potential novel targets for the treatment of inflammatory skin disorders. Human and mouse IL-36RN share 90% sequence identity.
  • This recombinant protein can be used for biological assays. For research use only.