Human Recombinant Lacritin (from E. coli)
Supplier: ProSci
PRSI91-130EA 455 EUR
PRSI91-130
Human Recombinant Lacritin (from E. coli)
Proteins and Peptides
Human Lacritin is highly expressed in the lacrimal gland, localizes primarily to secretory granules and secretory fluid. Lacritin modulates lacrimal acinar cell secretion, promotes ductal cell proliferation, and stimulates signaling through tyrosine phosphorylation and release of calcium.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag