Human Recombinant OTUB2 (from E. coli)
Supplier: ProSci
PRSI91-222EA 610 EUR
PRSI91-222
Human Recombinant OTUB2 (from E. coli)
Proteins and Peptides
Ubiquitin Thioesterase OTUB2 belongs to the peptidase C65 family, functions as a hydrolase that can remove conjugated ubiquitin from proteins in vitro, may plays an important regulatory role at the level of protein turnover by preventing degradation. OTUB2 is expressed at higher levels in the brain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag