Human Recombinant Visinin-like 1 (from E. coli)
Supplier: ProSci
PRSI91-245EA 580 EUR
PRSI91-245
Human Recombinant Visinin-like 1 (from E. coli)
Proteins and Peptides
Visinin-Like Protein 1 (VILIP) is a a member of the Visinin/Recoverin subfamily of neuronal calcium sensor proteins. VILIP is strongly expressed in the Granule Cells of the Cerebellum where it associates with membranes in a Calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of Adenylyl Cyclase. It has been shown that VILIP regulates the inhibition of rhodopsin phosphorylation in a Calcium-dependent manner in vitro.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag