Human Recombinant ZMYND19 (from E. coli)
Supplier: ProSci
PRSI91-260EA 580 EUR
PRSI91-260
Human Recombinant ZMYND19 (from E. coli)
Proteins and Peptides
Human Zinc Finger MYND Domain-Containing Protein 19 (ZMYND19) is a protein that contains 1 MYND-Type Zinc Finger. ZMYND19 can be expressed by the brain, testis, placenta, heart, liver, skeletal muscle, kidney, and stomach. ZMYND19 interacts with GPR24/MCH-R1. It binds to the C terminus of Melanin-Concentrating Hormone Receptor-1 and the N Termini of alpha-Tubulin. ZMYND19 may be involved as a regulatory molecule in GPR24/MCH-R1 signaling.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag