Human Recombinant CD47 (from Cells)
Supplier: ProSci
PRSI91-298EA 370 EUR
PRSI91-298
Human Recombinant CD47 (from Cells)
Proteins and Peptides
CD47 is involved in the increase in intracellular calcium concentration that occurs upon cell adhesion to extracellular matrix. The protein is also a receptor for the C-terminal cell-binding domain of thrombospondin, and it may play a role in membrane transport and signal transduction. This protein has broad tissue distribution, and is reduced in expression on Rh erythrocytes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag